Diethyl (bromodifluoromethyl)phosphonate 97%

Supplier: Ambeed
Warning

A196840-1000G A196840-100G A196840-10G A196840-500G A196840-25G A196840-5G A196840-1G
103816-866EA 9 USD
103816-866 103847-480 76800-196 76800-198 76800-200 76800-202 77085-454 77085-456 77302-528
Diethyl (bromodifluoromethyl)phosphonate 97%
Diethyl (bromodifluoromethyl)phosphonate

Precaución: for Research Use Only

Formula: C₅H₁₀BrF₂O₃P
Boiling Pt: 195.7 °C (760 mm Hg)
Flash Pt: 72.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00069118
CAS Number: 65094-22-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR