Diethyl ((phenylsulfonyl)methyl)phosphonate 97%

Supplier: Ambeed
Warning

A252733-10G A252733-250MG A252733-1G A252733-25G A252733-100G
103812-056EA 11 USD
103812-056 103851-904 77014-024 77014-026 77091-326 77091-328 77683-758
Diethyl ((phenylsulfonyl)methyl)phosphonate 97%
Diethyl ((phenylsulfonyl)methyl)phosphonate

Precaución: For research use only.

Formula: C₁₁H₁₇O₅PS
Boiling Pt: 392.3 °C (760 mm Hg)
Melting Pt: 145…148 °C
Flash Pt: 191.1 °C
Storage Temperature: Ambient
MDL Number: MFCD03453041
CAS Number: 56069-39-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR