Diethyl ((5-(3-fluorophenyl)pyridin-2-yl)methyl)phosphonate 97%

Supplier: Ambeed
Warning

A181435-100MG A181435-250MG
103807-606EA 75 USD
103807-606 103845-978 77163-898 77163-900
Diethyl ((5-(3-fluorophenyl)pyridin-2-yl)methyl)phosphonate 97%
Diethyl ((5-(3-fluorophenyl)pyridin-2-yl)methyl)phosphonate

Precaución: Inert atmosphere, room temperature

Formula: C₁₆H₁₉FNO₃P
Boiling Pt: 445.2±45.0 °C (760 mm Hg)
Flash Pt: 223.1 °C
Storage Temperature: Ambient
MDL Number: MFCD22376876
CAS Number: 380894-77-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR