Boc-8-amino-3,6-dioxaoctanoic acid DCHA 98%

Supplier: Ambeed
Warning

Synonyms: Boc-AEEA.DCHA, Dicyclohexylamine 2,2-dimethyl-4-oxo-3,8,11-trioxa-5-azatridecan-13-oate

A452554-100MG A452554-250MG A452554-1G
103827-802EA 76 USD
103827-802 103838-872 76979-504 76979-506 77307-366
Boc-8-amino-3,6-dioxaoctanoic acid DCHA 98%
Boc-8-amino-3,6-dioxaoctanoic acid DCHA

Precaución: for Research Use Only

Formula: C₂₃H₄₄N₂O₆
MW: 444.62 g/mol
Boiling Pt: 425.7 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD06796877
CAS Number: 560088-79-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR