Dicyclohexyl(o-tolyl)phosphine 97%, Ambeed.Inc

Supplier: Ambeed
Warning

A103595-1G A103595-250MG
103796-574EA 18 USD
103796-574 103822-280 76982-306 77309-158 77682-949
Dicyclohexyl(o-tolyl)phosphine 97%, Ambeed.Inc
Dicyclohexyl(o-tolyl)phosphine

Precaución: For research use only.

Formula: C₁₉H₂₉P
Boiling Pt: 409.1 °C (760 mm Hg)
Melting Pt: 90…93 °C
Flash Pt: 212.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08276773
CAS Number: 173593-25-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR