2-(Dicyclohexylphosphino)-2'-methylbiphenyl 97%

Supplier: Ambeed
Warning

Synonyms: Dicyclohexyl(2'-methyl-[1,1'-biphenyl]-2-yl)phosphine, 2-Dicyclohexylphosphino-2'-methyl-)-1,1'-biphenyl, MePhos, 2-Dicyclohexylphosphino-2'-methylbiphenyl

A163358-100G A163358-10G A163358-250MG A163358-1G A163358-25G A163358-5G
103805-718EA 9 USD
103805-718 103843-978 77235-936 77235-938 77235-940 77320-064 77320-066 77320-068
2-(Dicyclohexylphosphino)-2'-methylbiphenyl 97%
2-(Dicyclohexylphosphino)-2'-methylbiphenyl

Precaución: for Research Use Only

Formula: C₂₅H₃₃P
MW: 364.513 g/mol
Boiling Pt: 499.3 °C (760 mm Hg)
Melting Pt: 105…109 °C
Flash Pt: 271.5 °C
Storage Temperature: Ambient
MDL Number: MFCD03094577
CAS Number: 251320-86-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR