Dicyclohexyl(2',6'-dimethoxy[1,1'-biphenyl]-2-yl)phosphine 98%

Supplier: Ambeed
Warning

Synonyms: 2-Dicyclohexylphosphino-2’,6’-dimethoxybiphenyl, SPhos, Dicyclohexyl(2',6'-dimethoxy-[1,1'-biphenyl]-2-yl)phosphine, 2-Dicyclohexylphosphino-2',6'-dimethoxybiphenyl, 2-Dicyclohexylphosphino-2',6'-dimethoxy-1,1'-biphenyl

A117430-100G A117430-10G A117430-25G A117430-500G A117430-1G A117430-5G
103798-886EA 13 USD
103798-886 103824-770 76810-252 76810-254 76810-256 76810-258 77297-540 77311-554
Dicyclohexyl(2',6'-dimethoxy[1,1'-biphenyl]-2-yl)phosphine 98%
Dicyclohexyl(2',6'-dimethoxy[1,1'-biphenyl]-2-yl)phosphine

Precaución: for Research Use Only

Formula: C₂₆H₃₅O₂P
MW: 410.53 g/mol
Boiling Pt: 513.3 °C (760 mm Hg)
Melting Pt: 164…166 °C
Storage Temperature: Ambient
MDL Number: MFCD05861611
CAS Number: 657408-07-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR