Bis(tricyclohexylphosphine)palladium(II) dichloride 98%

Supplier: Ambeed
Warning

Synonyms: trans-Dichlorobis(tricyclohexylphosphine)palladium(II), Dichlorobis(tricyclohexylphosphine)palladium(II)

A113862-1G A113862-10G A113862-250MG
103798-306EA 77 USD
103798-306 103824-148 77310-964 77623-948 77623-950
Bis(tricyclohexylphosphine)palladium(II) dichloride 98%
Bis(tricyclohexylphosphine)palladium(II) dichloride

Precaución: for Research Use Only

Formula: C₃₆H₆₆Cl₂P₂Pd
MW: 738.19 g/mol
Boiling Pt: 383.4 °C (760 mm Hg)
Melting Pt: 270 °C
Flash Pt: 195.6 °C
Storage Temperature: Freezer
MDL Number: MFCD00191830
CAS Number: 29934-17-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR