Dichloro[9,9-dimethyl-4,5-bis(diphenylphosphino)xanthene]palladium(II) 98%

Supplier: Ambeed
Warning

Synonyms: [4,5-Bis(diphenylphosphino)-9,9-dimethylxanthene]palladium(II) Dichloride, [4,5-Bis(diphenylphosphino)-9,9-dimethylxanthene]dichloropalladium(II)

A262770-1G A262770-5G A262770-250MG A262770-25G
103812-580EA 28 USD
103812-580 103852-476 77092-102 77092-104 77166-228 77166-230
Dichloro[9,9-dimethyl-4,5-bis(diphenylphosphino)xanthene]palladium(II) 98%
Dichloro[9,9-dimethyl-4,5-bis(diphenylphosphino)xanthene]palladium(II)

Precaución: for Research Use Only

Formula: C₃₉H₃₂Cl₂OP₂Pd
MW: 755.95 g/mol
Melting Pt: 280…287 °C
Storage Temperature: Refrigerator
MDL Number: MFCD14155707
CAS Number: 205319-10-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR