(Pentamethylcyclopentadienyl)rhodium(III) dichloride dimer 98%

Supplier: Ambeed
Warning

Synonyms: Dichloro(pentamethylcyclopentadienyl)rhodium (III) dimer, [Rh(Cp-)Cl2]2, Dichloro(pentamethylcyclopentadienyl)rhodium(III) dimer, Dichloro(pentamethylcyclopentadienyl)rhodium(III) Dimer

A387123-250MG A387123-100MG
103826-814EA 257 USD
103826-814 103837-856 76803-786 77572-158
(Pentamethylcyclopentadienyl)rhodium(III) dichloride dimer 98%
(Pentamethylcyclopentadienyl)rhodium(III) dichloride dimer

Precaución: for Research Use Only

Formula: C₂₀H₃₀Cl₄Rh₂
MW: 618.07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00061552
CAS Number: 12354-85-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR