(Pentamethylcyclopentadienyl)iridium(III) dichloride dimer 97%

Supplier: Ambeed
Danger

Synonyms: Dichloro(pentamethylcyclopentadienyl)iridium(III) Dimer, Dichloro(pentamethylcyclopentadienyl)iridium(III) dimer, [Ir(Cp-)Cl2]2, (Pentamethylcyclopentadienyl)iridium (III) chloride dimer

A360791-250MG A360791-5G A360791-100MG
103826-270EA 198 USD
103826-270 103837-280 76803-458 76803-460 77601-600
(Pentamethylcyclopentadienyl)iridium(III) dichloride dimer 97%
(Pentamethylcyclopentadienyl)iridium(III) dichloride dimer

Precaución: for Research Use Only

Formula: C₂₀H₃₀Cl₄Ir₂
MW: 796.69 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00075435
CAS Number: 12354-84-6
UN: 1759
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR