Dichloro(1,5-cyclooctadiene)ruthenium(II), polymer 97%

Supplier: Ambeed
Warning

A181944-250MG A181944-25G A181944-1G A181944-5G
103807-670EA 31 USD
103807-670 103846-040 76799-750 76799-752 77083-566 77083-568
Dichloro(1,5-cyclooctadiene)ruthenium(II), polymer 97%
Dichloro(1,5-cyclooctadiene)ruthenium(II), polymer

Precaución: for Research Use Only

Formula: [RuCl₂(C₈H₁₂)]n
Melting Pt: 205 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00171304
CAS Number: 50982-12-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR