Dibenzyl diisopropylphosphoramidite 95%

Supplier: Ambeed
Warning

Synonyms: Dibenzyl N,N-diisopropylphosphoramidite

A179506-1G A179506-25G A179506-5G A179506-100G A179506-10G A179506-500G
103807-436EA 17 USD
103807-436 103845-784 77083-246 77083-248 77083-250 77111-360 77111-362 77111-364
Dibenzyl diisopropylphosphoramidite 95%
Dibenzyl diisopropylphosphoramidite

Precaución: for Research Use Only

Formula: C₂₀H₂₈NO₂P
MW: 345.43 g/mol
Boiling Pt: 130 °C (760 mm Hg)
Flash Pt: 69 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00191988
CAS Number: 108549-23-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR