Dibenzothiophene-5,5-dioxide 98%

Supplier: Ambeed
Warning

A364538-100G A364538-10G A364538-1G A364538-5G A364538-25G
103837-380EA 16 USD
103837-380 76724-642 76724-644 76724-646 76840-612 77306-756
Dibenzothiophene-5,5-dioxide 98%
Dibenzothiophene-5,5-dioxide

Precaución: for Research Use Only

Formula: C₁₂H₈O₂S
Boiling Pt: 422.2 °C (760 mm Hg)
Melting Pt: 231…233 °C
Flash Pt: 275.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00004970
CAS Number: 1016-05-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR