4-Dibenzofuranboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: Dibenzofuran-4-boronic acid, Dibenzo[b,d]furan-4-ylboronic acid

A265658-100G A265658-10G A265658-25G A265658-5G A265658-1G
103812-744EA 5 USD
103812-744 103852-640 76722-928 76722-930 77092-342 77092-344 77304-924
4-Dibenzofuranboronic acid 98%
4-Dibenzofuranboronic acid

Precaución: for Research Use Only

Formula: C₁₂H₉BO₃
Melting Pt: 286…291 °C
Flash Pt: 219 °C
Storage Temperature: Ambient
MDL Number: MFCD00092336
CAS Number: 100124-06-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR