Dibenzofuran-2-ylboronic acid 95%

Supplier: Ambeed
Warning

A266020-100G A266020-10G A266020-500G A266020-1G A266020-5G A266020-25G
103812-772EA 6 USD
103812-772 103852-668 76827-212 76827-214 76827-216 77092-362 77092-364 77304-942
Dibenzofuran-2-ylboronic acid 95%
Dibenzofuran-2-ylboronic acid

Precaución: for Research Use Only

Formula: C₁₂H₉BO₃
Boiling Pt: 439 °C (760 mm Hg)
Flash Pt: 219 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11183307
CAS Number: 402936-15-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR