1,1'-(Azodicarbonyl)dipiperidine 98%

Supplier: Ambeed
Danger

Synonyms: Diazene-1,2-diylbis(piperidin-1-ylmethanone), ADDP

A254768-100G A254768-10G A254768-500G A254768-1G A254768-25G A254768-5G
103812-166EA 5 USD
103812-166 103852-018 77031-638 77031-640 77031-642 77091-462 77091-464 77091-466
1,1'-(Azodicarbonyl)dipiperidine 98%
1,1'-(Azodicarbonyl)dipiperidine

Precaución: for Research Use Only

Formula: C₁₂H₂₀N₄O₂
MW: 252.32 g/mol
Melting Pt: 134…136 °C
Flash Pt: 175.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00010111
CAS Number: 10465-81-3
UN: 1325
ADR: 4.1,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR