1,1'-Carbonyldi(1H-1,2,4-triazole) 95%
Supplier: Ambeed
|
Warning
|
A105434-100G
A105434-10G
A105434-25G
A105434-500G
A105434-5G
103822-618EA
9
USD
103822-618
77188-754
77188-756
77188-758
77188-760
77309-488
1,1'-Carbonyldi(1H-1,2,4-triazole) 95%
1,1'-Carbonyldi(1H-1,2,4-triazole)
Precaución: for Research Use Only
Formula:
C₅H₄N₆O Melting Pt: 145…150 °C Flash Pt: 213.3 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00043399 CAS Number: 41864-22-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...