D-Homophenylalanine ethyl ester hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: H-D-HoPhe-OEt·HCl

A184650-1G A184650-5G
103815-770EA 8 USD
103815-770 103846-334 77301-906 77301-908
D-Homophenylalanine ethyl ester hydrochloride 97%
D-Homophenylalanine ethyl ester hydrochloride

Precaución: for Research Use Only

Formula: C₁₂H₁₈ClNO₂
MW: 243.731 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03095466
CAS Number: 90940-54-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR