D-(+)-Glucurono-6,3-lactone 98%

Supplier: Ambeed
Warning

Synonyms: (R)-2-((2S,3R,4S)-3,4-Dihydroxy-5-oxotetrahydrofuran-2-yl)-2-hydroxyacetaldehyde, D-(+)-Glucuronolactone

A695148-1000G A695148-25G A695148-100G A695148-10G A695148-500G
103810-518EA 77 USD
103810-518 76844-820 76844-822 77005-948 77005-950 77005-952
D-(+)-Glucurono-6,3-lactone 98%
D(+)-Glucurono-6,3-lactone

Precaución: for Research Use Only

Formula: C₆H₈O₆
MW: 176.12 g/mol
Boiling Pt: 488.9 °C (760 mm Hg)
Flash Pt: 214.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00135622
CAS Number: 32449-92-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR