Dexrazoxane 98%

Supplier: Ambeed
Warning

Synonyms: (S)-4,4'-(Propane-1,2-diyl)bis(piperazine-2,6-dione), 4,4'-[(1S)-1-Methyl-1,2-ethanediyl]bis-2,6-piperazinedione, (S)-1,2-Bis(3,5-dioxo-1-piperazinyl)propane

A170417-100MG A170417-10MG A170417-1MG A170417-250MG A170417-50MG A170417-1G A170417-5G
103806-488EA 95 USD
103806-488 103844-764 76690-154 76690-156 76690-158 76690-160 76690-162 77301-048 77301-050
Dexrazoxane 98%
Dexrazoxane

Precaución: for Research Use Only

Formula: C₁₁H₁₆N₄O₄
MW: 268.27 g/mol
Boiling Pt: 531.5 °C (760 mm Hg)
Melting Pt: 194…196 °C
Storage Temperature: Ambient
MDL Number: MFCD00866449
CAS Number: 24584-09-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR