4-[(2-Tetrahydropyranyl)oxy]phenol 98%

Supplier: Ambeed
Warning

Synonyms: Deoxyarbutin, 4-((Tetrahydro-2H-pyran-2-yl)oxy)phenol, 4-(Tetrahydropyran-2-yloxy)phenol

A243777-100G A243777-10G A243777-1G A243777-25G A243777-5G A243777-100MG
103811-550EA 11 USD
103811-550 103851-374 76815-482 76815-484 77090-578 77090-580 77090-582 77672-182
4-[(2-Tetrahydropyranyl)oxy]phenol 98%
4-[(2-Tetrahydropyranyl)oxy]phenol

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₃
MW: 194.227 g/mol
Boiling Pt: 349.8 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD09970981
CAS Number: 53936-56-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR