Decaethylene glycol 97%

Supplier: Ambeed
Warning

A287939-250MG A287939-1G A287939-25G
103813-776EA 15 USD
103813-776 103834-918 76693-182 76837-468 76837-470
Decaethylene glycol 97%
Decaethylene glycol

Precaución: for Research Use Only

Formula: C₂₀H₄₂O₁₁
Boiling Pt: 539.5 °C (760 mm Hg)
Melting Pt: 28 °C
Flash Pt: 280.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00698693
CAS Number: 5579-66-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR