D-(-)-Cysteine 97%

Supplier: Ambeed
Warning

Synonyms: (S)-2-Amino-3-mercaptopropanoic acid, H-D-Cys-OH, (S)-(-)-Cysteine

A110205-100G A110205-10G A110205-25G A110205-500G A110205-5G
103797-640EA 9 USD
103797-640 103823-448 77107-890 77107-892 77107-894 77107-896 77310-246
D-(-)-Cysteine 97%
D-(-)-Cysteine

Precaución: for Research Use Only

Formula: C₃H₇NO₂S
MW: 121.16
Melting Pt: 230 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00066461
CAS Number: 921-01-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR