Dansyl chloride 97%

Supplier: Ambeed
Danger

Synonyms: 5-(Dimethylamino)naphthalene-1-sulphonyl chloride

A427499-10G A427499-1MG A427499-250MG A427499-1G A427499-5G
103827-486EA 14 USD
103827-486 103838-540 76725-294 76725-296 76725-298 76842-196 76842-198
Dansyl chloride 97%
Dansyl chloride

Precaución: for Research Use Only

Formula: C₁₂H₁₂ClNO₂S
MW: 269.75 g/mol
Boiling Pt: 371.3 °C (760 mm Hg)
Melting Pt: 70…73 °C
Flash Pt: 178.4 °C
Storage Temperature: Freezer
MDL Number: MFCD00003985
CAS Number: 605-65-2
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR