Danofloxacin mesylate 98%

Supplier: Ambeed
Warning

Synonyms: CP-76136-27, 1-Cyclopropyl-6-fluoro-7-((1S,4S)-5-methyl-2,5-diazabicyclo[2.2.1]heptan-2-yl)-4-oxo-1,4-dihydroquinoline-3-carboxylic acid methanesulphonic acid salt

A302286-100MG A302286-25G A302286-50MG A302286-5G A302286-1G A302286-10MG
103814-334EA 10 USD
103814-334 103835-490 76693-428 76693-430 76693-432 76838-176 77305-850 77626-532
Danofloxacin mesylate 98%
Danofloxacin mesylate

Precaución: for Research Use Only

Formula: C₂₀H₂₄FN₃O₆S
MW: 453.48 g/mol
Boiling Pt: 569.3 °C (760 mm Hg)
Melting Pt: 337…339 °C
Flash Pt: 298.1 °C
Storage Temperature: Freezer
MDL Number: MFCD00673687
CAS Number: 119478-55-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR