D-Alanine ethyl ester hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: H-D-Ala-OEt·HCl, H-D-Ala-OEt hydrochloride

A178108-100G A178108-10G A178108-5G A178108-25G
103809-116EA 455 USD
103809-116 76813-446 76813-448 76813-450 77301-540
D-Alanine ethyl ester hydrochloride 98%
D-Alanine ethyl ester hydrochloride

Precaución: for Research Use Only

Formula: C₅H₁₂ClNO₂
MW: 153.61 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00190725
CAS Number: 6331-09-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR