Cyclopropyltriphenylphosphonium bromide 95%

Supplier: Ambeed
Warning

A195753-100G A195753-10G A195753-5G A195753-25G
103847-392EA 9 USD
103847-392 76691-060 76691-062 77085-342 77302-480
Cyclopropyltriphenylphosphonium bromide 95%
Cyclopropyltriphenylphosphonium bromide

Precaución: for Research Use Only

Formula: C₂₁H₂₀BrP
MW: 383.27 g/mol
Melting Pt: 178…181 °C
Storage Temperature: Ambient
MDL Number: MFCD00011872
CAS Number: 14114-05-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR