Cyclopropylhydrazine hydrochloride 95%

Supplier: Ambeed
Warning

A200624-1G A200624-250MG
103817-226EA 18 USD
103817-226 103847-858 77085-918 77883-459
Cyclopropylhydrazine hydrochloride 95%
Cyclopropylhydrazine hydrochloride

Precaución: for Research Use Only

Formula: C₃H₉ClN₂
Storage Temperature: Freezer
MDL Number: MFCD16658753
CAS Number: 213764-25-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR