Cyclopropylboronic acid 96% (by GC)
Supplier: Ambeed
|
Warning
|
A444176-100G
A444176-10G
103827-706EA
11
USD
103827-706
103838-774
76979-410
76979-412
Cyclopropylboronic acid 96% (by GC)
Cyclopropylboronic acid
Precaución: for Research Use Only
Formula:
C₃H₇BO₂ MW: 85.90 g/mol Boiling Pt: 205.1 °C (760 mm Hg) Melting Pt: 90…95 °C Flash Pt: 77.9 °C Storage Temperature: Freezer |
MDL Number:
MFCD04038750 CAS Number: 411235-57-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...