Cyclopropanesulfonamide 98%

Supplier: Ambeed
Warning

A115155-100G A115155-10G A115155-25G A115155-500G A115155-1G A115155-5G
103798-524EA 9 USD
103798-524 103824-370 76995-454 76995-456 76995-458 76995-460 77297-348 77297-350
Cyclopropanesulfonamide 98%
Cyclopropanesulphonamide

Precaución: for Research Use Only

Formula: C₃H₇NO₂S
Boiling Pt: 263 °C (760 mm Hg)
Melting Pt: 95…98 °C
Storage Temperature: Ambient
MDL Number: MFCD08705286
CAS Number: 154350-29-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR