Cyclopentanone-2-carbonitrile 95%
Supplier: Ambeed
|
Warning
|
A227664-1G
A227664-25G
A227664-5G
A227664-100G
A227664-10G
A227664-250MG
103819-334EA
10
USD
103819-334
103850-242
77089-018
77089-020
77089-022
77139-964
77139-966
77139-968
Cyclopentanone-2-carbonitrile 95%
Cyclopentanone-2-carbonitrile
Precaución: for Research Use Only
Formula:
C₆H₇NO Boiling Pt: 258 °C (760 mm Hg) Flash Pt: 109.9 °C Storage Temperature: Ambient |
MDL Number:
MFCD00154838 CAS Number: 2941-29-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...