1,3-Cyclopentanedione 97%

Supplier: Ambeed
Warning

A124196-100G A124196-10G A124196-25G A124196-1G A124196-5G A124196-500G
103800-500EA 5 USD
103800-500 77189-818 77189-820 77189-822 77297-976 77312-632 77745-911
1,3-Cyclopentanedione 97%
1,3-Cyclopentanedione

Precaución: for Research Use Only

Formula: C₅H₆O₂
Boiling Pt: 221.8 °C (760 mm Hg)
Melting Pt: 149…151 °C
Flash Pt: 80.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00001405
CAS Number: 3859-41-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR