3-Cyclopentene-1-carboxylic acid 97%

Supplier: Ambeed
Danger

Synonyms: cyclopent-3-ene-1-carboxylic acid, Cyclopent-3-enecarboxylic acid

A117980-10G A117980-25G A117980-1G A117980-5G
103798-970EA 5 USD
103798-970 103824-860 77026-230 77026-232 77297-568 77311-648
3-Cyclopentene-1-carboxylic acid 97%
3-Cyclopentene-1-carboxylic acid

Precaución: for Research Use Only

Formula: C₆H₈O₂
MW: 112.13 g/mol
Boiling Pt: 227.3 °C (760 mm Hg)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00101764
CAS Number: 7686-77-3
UN: 3265
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR