4-Aminocyclopentene hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: Cyclopent-3-enamine hydrochloride

A119130-100MG A119130-250MG A119130-25G A119130-1G A119130-5G
103799-168EA 27 USD
103799-168 77108-456 77108-458 77108-460 77311-842 77311-844
4-Aminocyclopentene hydrochloride 97%
4-Aminocyclopentene hydrochloride

Precaución: for Research Use Only

Formula: C₅H₁₀ClN
MW: 119.6 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01630740
CAS Number: 91469-55-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR