Cyclohexylboronic acid 98%
Supplier: Ambeed
|
Warning
|
A214406-25G
A214406-5G
A214406-10G
A214406-1G
A214406-100G
103818-398EA
10
USD
103818-398
103849-104
77087-620
77087-622
77152-550
77152-552
77612-106
Cyclohexylboronic acid 98%
Cyclohexylboronic acid
Precaución: for Research Use Only
Formula:
C₆H₁₃BO₂ Boiling Pt: 252.4 °C (760 mm Hg) Melting Pt: 122…123 °C Flash Pt: 106.4 °C Storage Temperature: Freezer |
MDL Number:
MFCD01311824 CAS Number: 4441-56-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...