Cyclohexanecarboxylic acid 98%

Supplier: Ambeed
Warning

A181341-100G A181341-500G A181341-1000G A181341-25G
103809-156EA 35 USD
103809-156 76824-932 76824-934 77083-496 77301-716
Cyclohexanecarboxylic acid 98%
Cyclohexanecarboxylic acid

Precaución: for Research Use Only

Formula: C₇H₁₂O₂
Boiling Pt: 232…233 °C (760 mm Hg)
Melting Pt: 29…31 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00001461
CAS Number: 98-89-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR