1,3-Cyclohexanedione 97%

Supplier: Ambeed
Warning

Synonyms: Cyclohexane-1,3-dione

A213419-1000G A213419-100G A213419-10G A213419-500G A213419-25G
103809-484EA 69 USD
103809-484 77087-520 77097-764 77097-766 77097-768 77303-286
1,3-Cyclohexanedione 97%
1,3-Cyclohexanedione

Precaución: for Research Use Only

Formula: C₆H₈O₂
MW: 112.13 g/mol
Boiling Pt: 235.1 °C (760 mm Hg)
Melting Pt: 101…105 °C
Flash Pt: 85.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001585
CAS Number: 504-02-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR