1-Cyclohexenylboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: Cyclohex-1-en-1-ylboronic acid

A275978-100MG A275978-10G
103813-284EA 7 USD
103813-284 103853-212 77166-556 77550-702
1-Cyclohexenylboronic acid 97%
1-Cyclohexenylboronic acid

Precaución: for Research Use Only

Formula: C₆H₁₁BO₂
MW: 125.96 g/mol
Boiling Pt: 261.9 °C (760 mm Hg)
Melting Pt: 165 °C
Flash Pt: 112.2 °C
Storage Temperature: Freezer
MDL Number: MFCD02179497
CAS Number: 89490-05-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR