Cycloheptanecarboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: Cycloheptane carboxylic acid

A102010-1G A102010-5G
103796-252EA 14 USD
103796-252 103821-952 77308-834 77308-836
Cycloheptanecarboxylic acid 98%
Cycloheptanecarboxylic acid

Precaución: for Research Use Only

Formula: C₈H₁₄O₂
MW: 142.2 g/mol
Boiling Pt: 135…138 °C (760 mm Hg)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00004152
CAS Number: 1460-16-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR