Copper(II) stearate 98%

Supplier: Ambeed
Warning

A322268-100G A322268-10G A322268-500G A322268-5G A322268-1000G A322268-25G
103810-110EA 93 USD
103810-110 76693-794 76693-796 76693-798 76693-800 76839-150 76839-152
Copper(II) stearate 98%
Copper(II) stearate

Precaución: for Research Use Only

Formula: C₃₆H₇₀CuO₄
MW: 630.498 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00050714
CAS Number: 660-60-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR