Copper(I)-2-thiophenecarboxylate 98%

Supplier: Ambeed
Warning

Synonyms: (2-Thiophenecarboxylato)copper(I), Copper(I) thiophene-2-carboxylate, 2-Thiophenecarboxylic Acid Copper(I) Salt, CuTC

A307966-1G A307966-25G A307966-5G A307966-10G A307966-500G A307966-100G
103814-540EA 5 USD
103814-540 103835-716 76838-468 76838-470 76838-472 77002-646 77002-648 77550-848
Copper(I)-2-thiophenecarboxylate 98%
Copper(I)-2-thiophenecarboxylate

Precaución: for Research Use Only

Formula: C₅H₃CuO₂S
Storage Temperature: Refrigerator
MDL Number: MFCD02183524
CAS Number: 68986-76-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR