Copper(I) iodide 99%

Supplier: Ambeed
Danger

Synonyms: Cuprous iodide

A531002-25G A531002-5G A531002-100G A531002-500G A531002-1000G
103839-676EA 8 USD
103839-676 77307-736 77307-738 77551-606 77551-608 77569-028
Copper(I) iodide 99%
Copper(I) iodide

Precaución: For research use only.

Formula: CuI
MW: 190.45 g/mol
Boiling Pt: 1290 °C (760 mm Hg)
Melting Pt: 605 °C
Flash Pt: 1290 °C
Storage Temperature: Ambient
MDL Number: MFCD00010978
CAS Number: 7681-65-4
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR