N-(5-Chloro-4-((4-chlorophenyl)(cyano)methyl)-2-methylphenyl)-2-hydroxy-3,5-diiodobenzamide 99+%

Supplier: Ambeed
Warning

Synonyms: Closantel

A220377-250MG A220377-25G A220377-25MG A220377-1G A220377-5G
103818-856EA 21 USD
103818-856 103849-738 76814-852 76814-854 76814-856 77303-554 77303-556
N-(5-Chloro-4-((4-chlorophenyl)(cyano)methyl)-2-methylphenyl)-2-hydroxy-3,5-diiodobenzamide 99+%
N-(5-Chloro-4-((4-chlorophenyl)(cyano)methyl)-2-methylphenyl)-2-hydroxy-3,5-diiodobenzamide

Precaución: for Research Use Only

Formula: C₂₂H₁₄Cl₂I₂N₂O₂
MW: 663.08 g/mol
Boiling Pt: 590.5 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD00661151
CAS Number: 57808-65-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR