Clorgyline hydrochloride 98%

Supplier: Ambeed
Danger

Synonyms: N-(3-(2,4-Dichlorophenoxy)propyl)-N-methylprop-2-yn-1-amine hydrochloride, N-​[3-​(2,​4-​dichlorophenoxy)propyl]-​N-​methyl-​2-​propyn-​1-​amine​ monohydrochloride, N-Methyl-N-propargyl-3-(2,4-dichlorophenoxy)propylamine hydrochloride, 2-Propynylamine, N-(3-(2,4-dichlorophenoxy)propyl)-N-methyl-, hydrochloride (7CI,8CI), Clorgyline HCl

A101686-100MG A101686-1MG A101686-5MG A101686-10MG
103791-944EA 41 USD
103791-944 103791-946 103796-090 103799-248 77093-512 77093-514 77093-516 77282-798
Clorgyline hydrochloride 98%
Clorgyline hydrochloride

Precaución: for Research Use Only

Formula: C₁₃H₁₅Cl₂NO·HCl
Storage Temperature: Ambient
MDL Number: MFCD00052012
CAS Number: 17780-75-5
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR