Clindamycin hydrochloride ≥98%

Supplier: Ambeed
Warning

Synonyms: Cleocin Hydrochloride, (2S,4R)-N-((1S,2S)-2-Chloro-1-((2R,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(methylthio)tetrahydro-2H-pyran-2-yl)propyl)-1-methyl-4-propylpyrrolidine-2-carboxamide hydrochloride, 7[S]-Chloro-7-deoxylincomycin, Clindamycin HCl

A203629-100MG A203629-1G A203629-5G A203629-100G A203629-10G A203629-500G
103848-132EA 26 USD
103848-132 77593-324 77593-326 77611-978 77685-516 77685-517 77900-765
Clindamycin hydrochloride ≥98%
Clindamycin hydrochloride

Precaución: Sealed in dry, at room temperature.

Formula: C₁₈H₃₃ClN₂O₅S·HCl
MW: 461.45 g/mol
Boiling Pt: 628.1 °C (760 mm Hg)
Melting Pt: 141 °C
Flash Pt: 333.6 °C
Storage Temperature: Ambient
MDL Number: MFCD07793327
CAS Number: 21462-39-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR