(3aR,6aS)-tert-Butyl 5-oxohexahydrocyclopenta[c]pyrrole-2(1H)-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: cis-tert-Butyl 5-oxohexahydrocyclopenta[c]pyrrole-2(1H)-carboxylate

A186623-1G A186623-25G A186623-5G A186623-100MG A186623-10G A186623-250MG
103815-948EA 15 USD
103815-948 103846-534 77084-202 77084-204 77084-206 77164-068 77164-070 77164-072
(3aR,6aS)-tert-Butyl 5-oxohexahydrocyclopenta[c]pyrrole-2(1H)-carboxylate 97%
(3aR,6aS)-tert-Butyl 5-oxohexahydrocyclopenta[c]pyrrole-2(1H)-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₁₉NO₃
MW: 225.284 g/mol
Boiling Pt: 325.8±35.0 °C (760 mm Hg)
Flash Pt: 150.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD18782907
CAS Number: 146231-54-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR