cis-Octahydropyrrolo[3,4-b]pyridine 95%

Supplier: Ambeed
Warning

Synonyms: cis-Octahydro-1H-pyrrolo[3,4-b]pyridine

A155462-100MG A155462-250MG A155462-5G A155462-1G
103804-762EA 68 USD
103804-762 76973-294 76973-296 76973-298 77318-682
cis-Octahydropyrrolo[3,4-b]pyridine 95%
cis-Octahydropyrrolo[3,4-b]pyridine

Precaución: for Research Use Only

Formula: C₇H₁₄N₂
MW: 126.2 g/mol
Boiling Pt: 198.5 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD04116298
CAS Number: 147459-51-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR