Methyl cis-3-hydroxycyclobutanecarboxylate 95%

Supplier: Ambeed
Warning

Synonyms: cis-Methyl 3-hydroxycyclobutanecarboxylate

A388632-250MG A388632-5G A388632-1G A388632-100G
103826-840EA 11 USD
103826-840 77168-158 77168-160 77306-986 77613-172
Methyl cis-3-hydroxycyclobutanecarboxylate 95%
Methyl cis-3-hydroxycyclobutanecarboxylate

Precaución: for Research Use Only

Formula: C₆H₁₀O₃
MW: 130.142 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18483124
CAS Number: 63485-50-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR