Diethyl-cis-cyclopropane-1,2-dicarboxylate 95%
Supplier: Ambeed
|
Warning
|
A142085-10G
A142085-250MG
A142085-1G
A142085-5G
103803-096EA
57
USD
103803-096
76972-646
76972-648
77299-266
77299-268
Diethyl-cis-cyclopropane-1,2-dicarboxylate 95%
Diethyl-cis-cyclopropane-1,2-dicarboxylate
Precaución: for Research Use Only
Formula:
C₉H₁₄O₄ Storage Temperature: Ambient |
MDL Number:
MFCD14525685 CAS Number: 710-43-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...